siRNA Details
| Gene Symbol: Srgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081011.2 | NP_001074480.2 | 15 | 2319 | ||
| GenBank mRNA | AK161932.1 | - | 4 | 223 | ||
| AY057900.1 | - | 6 | 308 | |||
| BC030457.1 | - | 14 | 1626 | |||
| BC151081.1 | - | 14 | 1937 | 9087994 | ||
| BC151082.1 | - | 14 | 1937 | 9087995 | ||
| BC158055.1 | - | 14 | 1935 | 9088163 | ||
| BC172152.1 | - | 14 | 1937 | 9054822 | ||

Srgap2
Genomic Map