siRNA Details
| Gene Symbol: Fpr-rs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008041.2 | NP_032067.2 | 1 | 603 | ||
| GenBank mRNA | BC115534.1 | - | 1 | 599 | ||
| BC115535.1 | - | 1 | 603 | |||
More Information
Other Species Targeted
Set Membership: |
|

Fpr-rs4
Genomic Map