siRNA Details
Gene Symbol: Fst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301373.1 | NP_001288302.1 | 3 | 733 | ||
NM_001301375.1 | NP_001288304.1 | 3 | 733 | |||
NM_008046.3 | NP_032072.1 | 3 | 733 | |||
GenBank mRNA | AK079916.1 | - | 3 | 734 | ||
AK083556.1 | - | 3 | 512 | |||
BC144926.1 | - | 3 | 557 | 9053474 | ||
BC145945.1 | - | 3 | 557 | 40131248 | ||
X83377.1 | - | 3 | 362 | |||
Z29532.1 | - | 3 | 368 |
