siRNA Details
| Gene Symbol: Fst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301373.1 | NP_001288302.1 | 4 | 970 | ||
| NM_001301375.1 | NP_001288304.1 | 4 | 970 | |||
| NM_008046.3 | NP_032072.1 | 4 | 970 | |||
| GenBank mRNA | AK079916.1 | - | 4 | 971 | ||
| BC144926.1 | - | 4 | 794 | 9053474 | ||
| BC145945.1 | - | 4 | 794 | 40131248 | ||
| X83377.1 | - | 4 | 599 | |||
| Z29532.1 | - | 4 | 605 | |||

Fst
Genomic Map