siRNA Details
| Gene Symbol: Fut9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010243.3 | NP_034373.1 | 3 | 1181 | ||
| GenBank mRNA | AB015426.1 | - | 3 | 785 | ||
| AK032177.1 | - | 3 | 1323 | |||
| AK033607.1 | - | 2 | 1336 | |||
| AK036183.1 | - | 3 | 984 | |||
| AK043410.1 | - | 3 | 1022 | |||
| AK047650.1 | - | 3 | 980 | |||
| AK049122.1 | - | 4 | 1262 | |||
| AK163871.1 | - | 3 | 981 | |||
| BC116874.1 | - | 3 | 877 | 40126193 | ||
| BC116876.1 | - | 3 | 877 | 40126195 | ||

Fut9
Genomic Map