siRNA Details
| Gene Symbol: Fxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113188.1 | NP_001106659.1 | 12 | 1304 | ||
| NM_001113189.1 | NP_001106660.1 | 13 | 1217 | |||
| NM_008053.2 | NP_032079.1 | 13 | 1217 | |||
| GenBank mRNA | AA080092.1 | - | 3 | 355 | ||
| AF124385.1 | - | 8 | 718 | |||
| AK049231.1 | - | 4 | 267 | |||
| AK198118.1 | - | 1 | 187 | |||
| BC019139.1 | - | 13 | 1198 | 5041635 | ||
| X90875.1 | - | 13 | 1196 | |||

Fxr1
Genomic Map