siRNA Details
| Gene Symbol: Fzd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162494.1 | NP_001155966.1 | 4 | 1084 | ||
| NM_008056.3 | NP_032082.3 | 4 | 1088 | |||
| GenBank mRNA | AK088399.1 | - | 4 | 953 | ||
| BC026150.1 | - | 4 | 1077 | 3983985 | ||
More Information
Set Membership: |
|

Fzd6
Genomic Map