siRNA Details
| Gene Symbol: Xrcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010247.2 | NP_034377.2 | 5 | 530 | ||
| GenBank mRNA | AB010282.1 | - | 5 | 471 | ||
| AF483500.1 | - | 4 | 410 | |||
| AF483501.1 | - | 4 | 410 | |||
| AK143570.1 | - | 5 | 505 | |||
| AK146392.1 | - | 5 | 449 | |||
| AK146394.1 | - | 5 | 449 | |||
| AK146396.1 | - | 5 | 497 | |||
| BC031422.1 | - | 5 | 479 | 3669715 | ||
| BC051085.1 | - | 5 | 1708 | |||
| M38700.1 | - | 4 | 372 | |||

Xrcc6
Genomic Map