siRNA Details
| Gene Symbol: G6pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008061.3 | NP_032087.2 | 5 | 1020 | ||
| GenBank mRNA | AK050279.1 | - | 5 | 1035 | ||
| AK052656.1 | - | 5 | 1035 | |||
| BC013448.1 | - | 5 | 1020 | 4237115 | ||
| U00445.1 | - | 5 | 1035 | |||

G6pc
Genomic Map