siRNA Details
| Gene Symbol: Slc37a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293630.1 | NP_001280559.1 | 8 | 1659 | ||
| NM_001293631.1 | NP_001280560.1 | 8 | 1372 | |||
| NM_008063.2 | NP_032089.2 | 7 | 1399 | |||
| GenBank mRNA | AF080469.1 | - | 7 | 1220 | ||
| AK003620.1 | - | 7 | 1222 | |||
| AK147849.1 | - | 8 | 1329 | |||
| AK155339.1 | - | 8 | 1656 | |||
| BC075655.1 | - | 7 | 1399 | 30542293 | ||

Slc37a4
Genomic Map