siRNA Details
| Gene Symbol: Slc37a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293630.1 | NP_001280559.1 | 9 | 1834 | ||
| NM_001293631.1 | NP_001280560.1 | 9 | 1547 | |||
| NM_008063.2 | NP_032089.2 | 8 | 1574 | |||
| GenBank mRNA | AF080469.1 | - | 8 | 1395 | ||
| AK003620.1 | - | 8 | 1397 | |||
| AK147849.1 | - | 9 | 1504 | |||
| AK155339.1 | - | 9 | 1831 | |||
| BC075655.1 | - | 8 | 1574 | 30542293 | ||

Slc37a4
Genomic Map