siRNA Details
| Gene Symbol: Gab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162477.1 | NP_001155949.1 | 4 | 736 | ||
| NM_010248.2 | NP_034378.2 | 4 | 736 | |||
| GenBank mRNA | AB018414.1 | - | 4 | 661 | ||
| AF104244.1 | - | 4 | 736 | |||
| BC103524.1 | - | 4 | 711 | 40039291 | ||
| BC103525.1 | - | 4 | 711 | 40039280 | ||
| BC103526.1 | - | 4 | 711 | 40039281 | ||
| BC103527.1 | - | 4 | 711 | 40039282 | ||

Gab2
Genomic Map