siRNA Details
Gene Symbol: Gabra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008066.3 | NP_032092.1 | 7, 8 | 996 | ||
GenBank mRNA | AK039055.1 | - | 7, 8 | 997 | ||
AK046953.1 | - | 2, 3 | 1130 | |||
AK048165.1 | - | 894 | ||||
BC115727.1 | - | 7, 8 | 918 | 40039253 | ||
M86567.1 | - | 7, 8 | 747 |
