siRNA Details
| Gene Symbol: Gabrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008069.4 | NP_032095.1 | 7, 8 | 830 | ||
| GenBank mRNA | BC130258.1 | - | 7, 8 | 834 | 40142758 | |
| BC157920.1 | - | 7, 8 | 896 | 9007350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrb1| Gene Symbol: Gabrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008069.4 | NP_032095.1 | 7, 8 | 830 | ||
| GenBank mRNA | BC130258.1 | - | 7, 8 | 834 | 40142758 | |
| BC157920.1 | - | 7, 8 | 896 | 9007350 | ||