siRNA Details
| Gene Symbol: Gabrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008070.3 | NP_032096.1 | 5 | 709 | ||
| GenBank mRNA | AK034069.1 | - | 4 | 324 | ||
| AK090279.1 | - | 5 | 710 | |||
| BC145973.1 | - | 5 | 683 | 40131276 | ||
| BC145975.1 | - | 5 | 683 | 40131278 | ||
| GU086167.1 | - | 4 | 282 | |||
| U14419.1 | - | 4 | 373 | |||

Gabrb2
Genomic Map