siRNA Details
| Gene Symbol: Gabre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017369.2 | NP_059065.2 | 6 | 1943 | ||
| GenBank mRNA | AF090376.1 | - | 2 | 64 | ||
| AF189263.1 | - | 6 | 1925 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabre| Gene Symbol: Gabre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017369.2 | NP_059065.2 | 6 | 1943 | ||
| GenBank mRNA | AF090376.1 | - | 2 | 64 | ||
| AF189263.1 | - | 6 | 1925 | |||