siRNA Details
| Gene Symbol: Gad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008078.2 | NP_032104.2 | 2, 3 | 468 | ||
| GenBank mRNA | AF326549.1 | - | 2, 3 | 132 | ||
| AF326550.1 | - | 2, 3 | 132 | |||
| AK136834.1 | - | 2, 3 | 168 | |||
| BC018380.1 | - | 2, 3 | 183 | 4482097 | ||
| BQ571205.1 | - | 2, 3 | 576 | |||
| D42051.1 | - | 2, 3 | 484 | |||
| KC134211.1 | - | 2, 3 | 132 | |||
| L16980.1 | - | 2, 3 | 204 | |||

Gad2
Genomic Map