siRNA Details
| Gene Symbol: Galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015736.2 | NP_056551.2 | 5 | 1298 | ||
| GenBank mRNA | AK019995.1 | - | 5 | 1297 | ||
| AK087219.1 | - | 1 | 364 | |||
| AK135489.1 | - | 5 | 1259 | |||
| BC043331.1 | - | 5 | 1154 | 5342768 | ||
| U70538.1 | - | 5 | 1142 | |||

Galnt3
Genomic Map