siRNA Details
| Gene Symbol: Usp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301628.1 | NP_001288557.1 | 19 | 2629 | ||
| NM_027604.4 | NP_081880.2 | 20 | 2716 | |||
| GenBank mRNA | AF468037.1 | - | 20 | 2620 | ||
| AK014891.1 | - | 14 | 1927 | |||
| AK122301.1 | - | 19 | 2544 | |||
| AK133852.1 | - | 19 | 2538 | |||
| AK145749.1 | - | 20 | 2620 | |||
| AK161469.1 | - | 20 | 2659 | |||
| AK161771.1 | - | 5 | 587 | |||
| AK163315.1 | - | 19 | 2560 | |||
| AK168755.1 | - | 19 | 2564 | |||
| BC003978.1 | - | 7 | 975 | |||
| BC050042.1 | - | 20 | 2632 | 30022307 | ||

Usp15
Genomic Map