siRNA Details
| Gene Symbol: Usp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301628.1 | NP_001288557.1 | 10 | 1466 | ||
| NM_027604.4 | NP_081880.2 | 11 | 1553 | |||
| GenBank mRNA | AF468037.1 | - | 11 | 1457 | ||
| AK014891.1 | - | 5 | 764 | |||
| AK052398.1 | - | 9 | 1282 | |||
| AK122301.1 | - | 10 | 1381 | |||
| AK133852.1 | - | 10 | 1375 | |||
| AK145749.1 | - | 11 | 1457 | |||
| AK161469.1 | - | 11 | 1496 | |||
| AK161957.1 | - | 10 | 1414 | |||
| AK163315.1 | - | 10 | 1395 | |||
| AK168755.1 | - | 10 | 1401 | |||
| BC050042.1 | - | 11 | 1469 | 30022307 | ||

Usp15
Genomic Map