siRNA Details
| Gene Symbol: Nr6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159548.1 | NP_001153020.1 | 6, 7 | 1048 | ||
| NM_001159549.1 | NP_001153021.1 | 6, 7 | 1045 | |||
| NM_010264.4 | NP_034394.1 | 8, 9 | 1356 | |||
| GenBank mRNA | AF390896.1 | - | 9, 10 | 1501 | ||
| CK638822.1 | - | 2, 3 | 445 | |||
| U09563.1 | - | 8, 9 | 1356 | |||
| U14666.1 | - | 8, 9 | 1315 | |||

Nr6a1
Genomic Map