siRNA Details
| Gene Symbol: Nr6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159548.1 | NP_001153020.1 | 2, 3 | 371 | ||
| NM_001159549.1 | NP_001153021.1 | 2, 3 | 371 | |||
| NM_010264.4 | NP_034394.1 | 4, 5 | 679 | |||
| GenBank mRNA | AF390896.1 | - | 5, 6 | 824 | ||
| AK141017.1 | - | 1, 2 | 3584 | |||
| CN537360.1 | - | 2, 3 | 347 | |||
| CN537407.1 | - | 2, 3 | 371 | |||
| CN538225.1 | - | 3, 4 | 350 | |||
| U09563.1 | - | 4, 5 | 679 | |||
| U14666.1 | - | 4, 5 | 638 | |||

Nr6a1
Genomic Map