siRNA Details
Gene Symbol: Opn1mw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008106.2 | NP_032132.1 | 2 | 340 | ||
GenBank mRNA | AF011389.1 | - | 2 | 326 | ||
AF190672.1 | - | 2 | 342 | |||
AK149287.1 | - | 2 | 341 | |||
BC014826.1 | - | 2 | 320 | 4507311 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
