siRNA Details
| Gene Symbol: Gdap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010269.3 | NP_034399.1 | 8 | 1097 | ||
| GenBank mRNA | AK004887.1 | - | 8 | 1043 | ||
| AK082958.1 | - | 8 | 1059 | |||
| AK146835.1 | - | 8 | 1087 | |||
| AK161122.1 | - | 8 | 996 | |||
| AK166799.1 | - | 8 | 1012 | |||
| AK166859.1 | - | 8 | 1011 | |||
| AK208025.1 | - | 1 | 48 | |||
| BC025070.1 | - | 8 | 1006 | 3710712 | ||
| Y17851.1 | - | 8 | 1094 | |||

Gdap2
Genomic Map