siRNA Details
| Gene Symbol: Gdf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008109.2 | NP_032135.2 | 2 | 1182 | ||
| GenBank mRNA | AB259648.1 | - | 2 | 880 | ||
| AK041168.1 | - | 1 | 136 | |||
| BC034546.1 | - | 2 | 1194 | 4190036 | ||
| U08337.1 | - | 2 | 1194 | |||

Gdf5
Genomic Map