siRNA Details
| Gene Symbol: Gdnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301332.1 | NP_001288261.1 | 2 | 1089 | ||
| NM_001301333.1 | NP_001288262.1 | 2 | 373 | |||
| NM_001301357.1 | NP_001288286.1 | 1 | 61 | |||
| NM_010275.3 | NP_034405.1 | 2 | 1089 | |||
| GenBank mRNA | BC119031.1 | - | 2 | 105 | 8733780 | |
| D88264.1 | - | 2 | 291 | |||
| U37459.1 | - | 2 | 126 | |||
| U75532.1 | - | 2 | 206 | |||

Gdnf
Genomic Map