siRNA Details
| Gene Symbol: Gfi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160406.1 | NP_001153878.1 | 3 | 319 | ||
| NM_008114.3 | NP_032140.1 | 3 | 319 | |||
| GenBank mRNA | AF017275.1 | - | 3 | 278 | ||
| AK157262.1 | - | 3 | 328 | |||
| BC052654.1 | - | 3 | 294 | 30039774 | ||
| BC132365.1 | - | 3 | 255 | 40130642 | ||
| BC132367.1 | - | 3 | 255 | 40130644 | ||
| BC145238.1 | - | 3 | 255 | 9053790 | ||
| W65842.1 | - | 3 | 318 | |||

Gfi1b
Genomic Map