siRNA Details
| Gene Symbol: Gfpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013529.3 | NP_038557.1 | 18 | 1969 | ||
| GenBank mRNA | AB016780.1 | - | 18 | 1935 | ||
| AK132821.1 | - | 17 | 1815 | |||
| BC031928.1 | - | 18 | 1937 | 4167189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfpt2| Gene Symbol: Gfpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013529.3 | NP_038557.1 | 18 | 1969 | ||
| GenBank mRNA | AB016780.1 | - | 18 | 1935 | ||
| AK132821.1 | - | 17 | 1815 | |||
| BC031928.1 | - | 18 | 1937 | 4167189 | ||