siRNA Details
| Gene Symbol: Gfpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013529.3 | NP_038557.1 | 10 | 955 | ||
| GenBank mRNA | AB016780.1 | - | 10 | 921 | ||
| AK132821.1 | - | 9 | 801 | |||
| BC031928.1 | - | 10 | 923 | 4167189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfpt2| Gene Symbol: Gfpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013529.3 | NP_038557.1 | 10 | 955 | ||
| GenBank mRNA | AB016780.1 | - | 10 | 921 | ||
| AK132821.1 | - | 9 | 801 | |||
| BC031928.1 | - | 10 | 923 | 4167189 | ||