siRNA Details
| Gene Symbol: Ggh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010281.2 | NP_034411.2 | 1, 2 | 226 | ||
| GenBank mRNA | AF051102.1 | - | 1, 2 | 194 | ||
| AK089996.1 | - | 1, 2 | 183 | |||
| BB610405.1 | - | 1, 2 | 114 | |||
| BC009809.1 | - | 1, 2 | 117 | 4223953 | ||
| BC150898.1 | - | 1, 2 | 163 | 9087809 | ||
| BE859353.1 | - | 1, 2 | 119 | |||

Ggh
Genomic Map