siRNA Details
| Gene Symbol: Ostm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172416.3 | NP_766004.1 | 5 | 887 | ||
| GenBank mRNA | AF533890.1 | - | 5 | 886 | ||
| AK004546.1 | - | 5 | 864 | |||
| AK044897.1 | - | 5 | 886 | |||
| AK050513.1 | - | 5 | 861 | |||
| AK053664.1 | - | 4 | 2526 | |||
| BC057635.1 | - | 5 | 825 | 4910872 | ||

Ostm1
Genomic Map