siRNA Details
| Gene Symbol: Gpam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008149.3 | NP_032175.2 | 13 | 1540 | ||
| GenBank mRNA | AK047295.1 | - | 13 | 1541 | ||
| AK137067.1 | - | 15 | 1821 | |||
| AK173202.1 | - | 13 | 1565 | |||
| BC019201.1 | - | 13 | 1545 | 5041120 | ||

Gpam
Genomic Map