siRNA Details
| Gene Symbol: Gpam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008149.3 | NP_032175.2 | 16 | 2023 | ||
| GenBank mRNA | AK047295.1 | - | 16 | 2024 | ||
| AK137067.1 | - | 18 | 2304 | |||
| AK173202.1 | - | 16 | 2048 | |||
| BC019201.1 | - | 16 | 2028 | 5041120 | ||
| BC082776.1 | - | 2 | 325 | |||

Gpam
Genomic Map