siRNA Details
| Gene Symbol: Gpam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008149.3 | NP_032175.2 | 5 | 480 | ||
| GenBank mRNA | AK047295.1 | - | 5 | 481 | ||
| AK137067.1 | - | 7 | 761 | |||
| AK173202.1 | - | 5 | 505 | |||
| BC019201.1 | - | 5 | 485 | 5041120 | ||

Gpam
Genomic Map