siRNA Details
| Gene Symbol: Grb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177629.1 | NP_001171100.1 | 12 | 1340 | ||
| NM_010345.4 | NP_034475.2 | 13 | 1665 | |||
| GenBank mRNA | AF022072.1 | - | 13 | 1639 | ||
| AK030727.1 | - | 13 | 1600 | |||
| AK032927.1 | - | 13 | 1646 | |||
| AK163841.1 | - | 13 | 1646 | |||
| AK169000.1 | - | 7 | 629 | |||
| BC016111.1 | - | 12 | 1340 | 4481345 | ||
| BC053842.1 | - | 13 | 1632 | |||

Grb10
Genomic Map