siRNA Details
| Gene Symbol: Grb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177629.1 | NP_001171100.1 | 6 | 730 | ||
| NM_010345.4 | NP_034475.2 | 7 | 1055 | |||
| GenBank mRNA | AF022072.1 | - | 7 | 1029 | ||
| AK030727.1 | - | 7 | 990 | |||
| AK032927.1 | - | 7 | 1036 | |||
| AK163841.1 | - | 7 | 1036 | |||
| AK169000.1 | - | 1 | 19 | |||
| BC016111.1 | - | 6 | 730 | 4481345 | ||
| BC053842.1 | - | 7 | 1022 | |||

Grb10
Genomic Map