siRNA Details
| Gene Symbol: Gpr162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013533.3 | NP_038561.1 | 3 | 1723 | ||
| GenBank mRNA | AK134941.1 | - | 3 | 1537 | ||
| AK144354.1 | - | 2 | 1492 | |||
| AK158847.1 | - | 3 | 2242 | |||
| AK171145.1 | - | 4 | 1907 | |||
| BC016413.1 | - | 3 | 780 | |||
| BC031437.1 | - | 3 | 1723 | 4527410 | ||

Gpr162
Genomic Map