siRNA Details
| Gene Symbol: Cdca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013538.5 | NP_038566.1 | 4 | 499 | ||
| GenBank mRNA | AK008606.1 | - | 4 | 452 | ||
| AK010409.1 | - | 4 | 440 | |||
| AK011313.1 | - | 4 | 472 | |||
| AK028159.1 | - | 4 | 441 | |||
| AK145537.1 | - | 4 | 441 | |||
| AK169792.1 | - | 4 | 447 | |||
| BC027172.1 | - | 3 | 1077 | |||

Cdca3
Genomic Map