siRNA Details
| Gene Symbol: Grid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008167.2 | NP_032193.1 | 6 | 882 | ||
| GenBank mRNA | AK036192.1 | - | 6 | 913 | ||
| AK141070.1 | - | 1734 | ||||
| BC139823.1 | - | 6 | 886 | 40141838 | ||
| D13266.1 | - | 6 | 882 | |||

Grid2
Genomic Map