siRNA Details
Gene Symbol: Grik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010348.3 | NP_034478.1 | 13 | 2447 | ||
NM_146072.4 | NP_666184.2 | 13 | 2447 | |||
GenBank mRNA | AK042635.1 | - | 13 | 2448 | ||
AK043542.1 | - | 13 | 2427 | |||
AK043737.1 | - | 13 | 2411 | |||
AK044250.1 | - | 13 | 2406 | |||
AK044420.1 | - | 13 | 2402 | |||
AK133667.1 | - | 13 | 2433 | |||
BC031822.1 | - | 12 | 1869 | 4501004 | ||
BC049275.1 | - | 12 | 1869 | 4501004 | ||
X66118.1 | - | 13 | 2433 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
