siRNA Details
Gene Symbol: Grik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010348.3 | NP_034478.1 | 6 | 1424 | ||
NM_146072.4 | NP_666184.2 | 6 | 1424 | |||
GenBank mRNA | AK042635.1 | - | 6 | 1425 | ||
AK043542.1 | - | 6 | 1404 | |||
AK043737.1 | - | 6 | 1388 | |||
AK044250.1 | - | 6 | 1383 | |||
AK044420.1 | - | 6 | 1378 | |||
AK046122.1 | - | 6 | 1431 | |||
AK048634.1 | - | 6 | 1395 | |||
AK133667.1 | - | 6 | 1410 | |||
BC031822.1 | - | 5 | 846 | 4501004 | ||
BC049275.1 | - | 5 | 846 | 4501004 | ||
X66118.1 | - | 6 | 1410 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001111117, NM_017241, NM_001111114, XM_006248053, XM_006248054, and others.
Panther Classification:
Gene Ontology Categories:
