siRNA Details
| Gene Symbol: Grn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008175.4 | NP_032201.2 | 12 | 1805 | ||
| GenBank mRNA | AK013678.1 | - | 12 | 1621 | ||
| AK018744.1 | - | 12 | 1646 | |||
| AK147932.1 | - | 12 | 1622 | |||
| AK149570.1 | - | 12 | 1623 | |||
| AK150205.1 | - | 12 | 1621 | |||
| AK150338.1 | - | 12 | 1610 | |||
| AK150420.1 | - | 12 | 1609 | |||
| AK150717.1 | - | 12 | 1621 | |||
| AK150746.1 | - | 12 | 1623 | |||
| AK150881.1 | - | 12 | 1623 | |||
| AK151007.1 | - | 12 | 1621 | |||
| AK151162.1 | - | 12 | 1623 | |||
| AK151343.1 | - | 12 | 1621 | |||
| AK151713.1 | - | 12 | 1621 | |||
| AK151756.1 | - | 12 | 1621 | |||
| AK152048.1 | - | 12 | 1621 | |||
| AK152087.1 | - | 12 | 1621 | |||
| AK152118.1 | - | 12 | 1610 | |||
| AK152213.1 | - | 12 | 1610 | |||
| AK152705.1 | - | 12 | 1621 | |||
| AK152765.1 | - | 12 | 1621 | |||
| AK152944.1 | - | 12 | 1610 | |||
| AK153183.1 | - | 12 | 1611 | |||
| AK153269.1 | - | 12 | 1620 | |||
| AK153289.1 | - | 12 | 1621 | |||
| AK153418.1 | - | 12 | 1621 | |||
| AK153470.1 | - | 12 | 1621 | |||
| AK154492.1 | - | 12 | 1621 | |||
| AK159397.1 | - | 12 | 1610 | |||
| AK159428.1 | - | 12 | 1610 | |||
| AK159557.1 | - | 12 | 1621 | |||
| AK159809.1 | - | 12 | 1621 | |||
| AK160022.1 | - | 12 | 1610 | |||
| AK160033.1 | - | 12 | 1621 | |||
| AK160074.1 | - | 12 | 1610 | |||
| AK185467.1 | - | 1 | 134 | |||
| BC037047.1 | - | 11 | 1602 | |||
| BC049943.1 | - | 12 | 1607 | |||
| BC129849.1 | - | 12 | 1605 | 40109630 | ||
| BC129850.1 | - | 12 | 1605 | 40109631 | ||
| M86736.1 | - | 12 | 1603 | |||
| X62321.1 | - | 11 | 1586 | |||

Grn
Genomic Map