siRNA Details
| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010369.3 | NP_034499.3 | 5, 6 | 332 | ||
| GenBank mRNA | AK145998.1 | - | 5, 6 | 333 | ||
| AK157851.1 | - | 5, 6 | 333 | |||
| AK162614.1 | - | 5, 6 | 317 | |||
| BC148192.1 | - | 5, 6 | 596 | 40045496 | ||

Gypa
Genomic Map