siRNA Details
| Gene Symbol: Cfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142706.1 | NP_001136178.1 | 12 | 2003 | ||
| NM_008198.2 | NP_032224.2 | 12 | 2003 | |||
| GenBank mRNA | AK098094.1 | - | 12 | 1663 | ||
| AK149531.1 | - | 12 | 1636 | |||
| AK149573.1 | - | 12 | 1662 | |||
| AK151220.1 | - | 6 | 622 | |||
| AK151618.1 | - | 6 | 622 | |||
| AK151974.1 | - | 6 | 622 | |||
| BC005451.1 | - | 12 | 1637 | 3591669 | ||
| K01496.1 | - | 7 | 743 | |||
| K01498.1 | - | 2 | 167 | |||
| M57890.1 | - | 12 | 1706 | |||

Cfb
Genomic Map