siRNA Details
| Gene Symbol: Mr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008209.4 | NP_032235.1 | 4 | 868 | ||
| GenBank mRNA | AF010448.1 | - | 4 | 779 | ||
| AF010452.1 | - | 4 | 594 | |||
| AK042119.1 | - | 4 | 709 | |||
| AK137171.1 | - | 4 | 709 | |||
| AK144040.1 | - | 4 | 813 | |||
| AK162712.1 | - | 4 | 825 | |||
| BC026137.1 | - | 4 | 896 | 4017492 | ||
| U94989.1 | - | 4 | 808 | |||

Mr1
Genomic Map