siRNA Details
| Gene Symbol: Mr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008209.4 | NP_032235.1 | 2 | 215 | ||
| GenBank mRNA | AF010448.1 | - | 2 | 126 | ||
| AK041171.1 | - | 2 | 249 | |||
| AK144040.1 | - | 2 | 240 | |||
| AK162712.1 | - | 2 | 173 | |||
| BC026137.1 | - | 2 | 243 | 4017492 | ||
| U94989.1 | - | 2 | 155 | |||

Mr1
Genomic Map