siRNA Details
| Gene Symbol: Hdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008230.6 | NP_032256.3 | 9 | 1136 | ||
| GenBank mRNA | AF109137.1 | - | 9 | 1046 | ||
| AK076384.1 | - | 8 | 3108 | |||
| AK088545.1 | - | 9 | 1123 | |||
| AK133455.1 | - | 9 | 1137 | |||
| AK150168.1 | - | 9 | 1124 | |||
| AK153104.1 | - | 9 | 1124 | |||
| BC052833.1 | - | 9 | 1121 | 30044098 | ||
| X57437.1 | - | 9 | 1101 | |||

Hdc
Genomic Map