siRNA Details
Gene Symbol: Hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008230.6 | NP_032256.3 | 6 | 734 | ||
GenBank mRNA | AF109137.1 | - | 6 | 644 | ||
AK076384.1 | - | 5 | 2706 | |||
AK088545.1 | - | 6 | 721 | |||
AK133455.1 | - | 6 | 735 | |||
AK150168.1 | - | 6 | 722 | |||
AK153104.1 | - | 6 | 722 | |||
BC052833.1 | - | 6 | 719 | 30044098 | ||
X57437.1 | - | 6 | 699 |
