siRNA Details
| Gene Symbol: Hebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013546.2 | NP_038574.2 | 3 | 427 | ||
| GenBank mRNA | AA022130.1 | - | 415 | |||
| AB013095.1 | - | 3 | 366 | |||
| AF117613.1 | - | 3 | 415 | |||
| AK008919.1 | - | 3 | 348 | |||
| BC012654.1 | - | 3 | 351 | 4216585 | ||
| BY242687.1 | - | 3 | 428 | |||
| CT010262.1 | - | 3 | 309 | |||

Hebp1
Genomic Map