siRNA Details
| Gene Symbol: Hells | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008234.3 | NP_032260.2 | 16 | 1874 | ||
| GenBank mRNA | AF155210.1 | - | 16 | 1874 | ||
| AK013266.1 | - | 16 | 1913 | |||
| AK147126.1 | - | 16 | 1905 | |||
| AK166636.1 | - | 16 | 1875 | |||
| BC020056.1 | - | 3 | 238 | |||
| BC090671.1 | - | 16 | 1918 | |||
| BC100394.1 | - | 16 | 1871 | 30480539 | ||
| U25691.2 | - | 16 | 1907 | |||

Hells
Genomic Map